Skip to Content
Merck
All Photos(1)

Documents

261017

Sigma-Aldrich

Mercury

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hg
CAS Number:
Molecular Weight:
200.59
EC Number:
MDL number:
UNSPSC Code:
12141726
PubChem Substance ID:
NACRES:
NA.55

vapor density

7 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 126 °C)

Assay

≥99.99% trace metals basis

form

liquid

resistivity

95.8 μΩ-cm, 20°C

bp

356.6 °C (lit.)
356.6 °C

mp

−38.87 °C (lit.)

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mercury (quicksilver) is a rare chemical element found most commonly in cinnabar (HgS). It is widely employed in the production of industrial chemicals like mercuric (I) and mercuric (II) compounds, because of two oxidation states (+1 and +2). Additionally, it is also used in the synthesis of coordination complexes of mercury as well as organomercury compounds. Mercury(II) salts are used in the preparation of industrially important chemical reagents such as Millon′s base, Nessler′s reagent (potassium tetraiodomercurate), one-dimensional polymer (salts of HgNH2+)n), etc.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sabrina Llop et al.
PloS one, 9(5), e97172-e97172 (2014-05-17)
The genetic background may influence methylmercury (MeHg) metabolism and neurotoxicity. ATP binding cassette (ABC) transporters actively transport various xenobiotics across biological membranes. To investigate the role of ABC polymorphisms as modifiers of prenatal exposure to MeHg. The study population consisted
M E Goodsite et al.
The Science of the total environment, 452-453, 196-207 (2013-03-20)
This review compares the reconstruction of atmospheric Hg deposition rates and historical trends over recent decades in the Arctic, inferred from Hg profiles in natural archives such as lake and marine sediments, peat bogs and glacial firn (permanent snowpack), against
Okechukwu S Ogah et al.
JACC. Heart failure, 2(3), 250-259 (2014-06-24)
The aim of this study was to determine the contemporary profile, clinical characteristics, and intrahospital outcomes of acute heart failure (AHF) in an African urban community. There are limited data on the current burden and characteristics of AHF in Nigerian
Ekim Seven et al.
Journal of hypertension, 32(7), 1488-1494 (2014-05-09)
The adipocytokines, leptin, adiponectin, and interleukin-6, which stimulate liver C-reactive protein (CRP) production, are regarded as potential candidate intermediates between adipose tissue and overweight-induced hypertension. We examined the associations between leptin, adiponectin, and CRP levels with both prevalent and 5-year
James P K Rooney
Toxicology, 234(3), 145-156 (2007-04-06)
Mercury has been a known as a toxic substance for centuries. Whilst the clinical features of acute mercury poisoning have been well described, chronic low dose exposure to mercury remains poorly characterised and its potential role in various chronic disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service