Skip to Content
Merck
All Photos(1)

Key Documents

247626

Sigma-Aldrich

Sulfur dioxide solution

ACS reagent, ≥6%

Synonym(s):

Sulfurous acid, Sulfurous anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO3
CAS Number:
Molecular Weight:
82.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
concentration:
≥6%
grade:
ACS reagent

grade

ACS reagent

Quality Level

vapor density

2.3 (vs air)

Assay

≥6% SO2 basis (by Na2S2O3, titration)

form

liquid

concentration

≥6%

ign. residue

≤0.005%

density

1.03 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤2 ppm

SMILES string

OS(O)=O

InChI

1S/H2O3S/c1-4(2)3/h(H2,1,2,3)

InChI key

LSNNMFCWUKXFEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfurous acid (H2SO3), a weak diprotic acid, is formed when sulfur dioxide (SO2) dissolves in water. Sulfurous acid is a complex mixture of various hydrates of SO2 and the ions H3O+, S2O52- and HSO3-. The predominant species present in the solution are pH dependent.

Application

Sulfurous acid can be used as a reducing agent in chemical synthesis.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Kharina et al.
Applied biochemistry and biotechnology, 179(2), 307-320 (2016-01-29)
Sulfurous acid was used for pretreatment of sugar beet pulp (SBP) in order to achieve high efficiency of both extraction of carbohydrates and subsequent enzymatic hydrolysis of the remaining solids. The main advantage of sulfurous acid usage as pretreatment agent
The thermodynamic ionization constants of sulfurous acid at 25?1.
Tartar HV and Garretson HH.
Journal of the American Chemical Society, 63(3), 808-816 (1941)
Miquel Pascual et al.
Journal of the science of food and agriculture, 95(15), 3067-3076 (2015-07-16)
Rainfed viticulture, mainly in semi-arid environments, is limited by environmental variability, particularly precipitation and its seasonal distribution, and soil water availability, thus ultimately determining the final quality of grape and wine. Studies on the feasibility of practices such as canopy
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 16(12), 3139-3143 (2006-04-20)
An inhibition study of the cytosolic carbonic anhydrase (CA, EC 4.2.1.1) isozyme VII (hCA VII) with anions has been conducted. Cyanate, cyanide, and hydrogensulfite were weak hCA VII inhibitors (K(I)s in the range of 7.3-15.2 mM). Cl- and HCO3- showed
Heng Yu et al.
Bioresource technology, 199, 188-193 (2015-09-06)
This study established a new more neutral magnesium bisulfate pretreatment (MBSP) using magnesium bisulfate as sulfonating agent for improving the enzymatic hydrolysis efficiency of corn stover. Using the MBSP with 5.21% magnesium bisulfate, 170°C and pH 5.2 for 60 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service