Skip to Content
Merck
All Photos(3)

Key Documents

223425

Sigma-Aldrich

Potassium oxalate monohydrate

ACS reagent, 99%

Synonym(s):

Ethanedioic acid, Oxalic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOK)2 · H2O
CAS Number:
Molecular Weight:
184.23
Beilstein:
3752576
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
crystals or chunks
solubility:
H2O: 10%
Pricing and availability is not currently available.

grade

ACS reagent

description

neutrality passes test

Assay

98.5-101.0% (ACS specification)
99%

form

crystals or chunks

impurities

H2SO4, passes test (darkened)
≤0.01% insolubles

pH

7.0-8.5 (25 °C, 50 g/L)

solubility

H2O: 10%

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%
NH4+: ≤0.002%
Na: ≤0.02%
heavy metals: ≤0.002% (by ICP)

SMILES string

O.[K+].[K+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium oxalate monohydrate (POM) is a potassium salt of oxalic acid. Its crystal structure has been investigated by three-dimensional neutron diffraction studies.[1]

Application

Potassium oxalate monohydrate may be used in the preparation of SBA-15 (Santa Barbara Amoprhous-15)- catalyst.[2]
Potassium oxalate monohydrate may be used in the synthesis of bis(2,2"-bipyridine) (oxalato-O,O′)nickel(ii) tetrahydrate, a mononuclear mixed-ligand complex.[3]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EPR and optical absorption studies of vanadyl ions in potassium oxalate monohydrate.
Kripal R, et al.
Physica B: Condensed Matter (Amsterdam, Netherlands), 392(1), 281-287 (2007)
Solid-state chemistry of nickel (II)-oxalate-amine systems. Synthesis, characterization and crystal structure of the bis (2, 2'-bipyridine)(oxalato-O, O') nickel (II) tetrahydrate.
Roman P, et al.
Polyhedron, 14(20), 2863-2869 null
Neutron diffraction refinement of the structure of potassium oxalate monohydrate.
Sequeira A, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 26(1), 77-80 (1970)
The influence of water on the dispersion of vanadia supported on silica SBA-15: A combined XPS and Raman study.
Hess C, et al.
The Journal of Physical Chemistry C, 111(26), 9471-9479 (2007)
L Sosvorová et al.
Physiological research, 63(4), 521-527 (2014-06-09)
Increased homocysteine levels in serum are typical features of neurodegenerative brain diseases including hydrocephalus. The most frequent therapeutic approach consists of the insertion of a shunt, connecting the brain ventricles to an alternative drainage site. To decide whether the patient

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service