Skip to Content
Merck
All Photos(3)

Documents

215198

Sigma-Aldrich

Sulfur

reagent grade, powder, purified by refining, -100 mesh particle size

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.55

grade

reagent grade

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

form

powder

autoignition temp.

450 °F

purified by

refining

resistivity

2E23 μΩ-cm, 20°C

particle size

-100 mesh

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application



  • Method comparison of HPLC-ninhydrin-photometry and UHPLC-PITC-tandem mass spectrometry for serum amino acid analyses in patients with complex congenital heart disease and controls.: This study compares two analytical methods, HPLC-ninhydrin-photometry and UHPLC-PITC-tandem mass spectrometry, for analyzing serum amino acids. It highlights the advantages of using Ion exchanger III in achieving accurate and reliable measurements in complex medical cases (Michel et al., 2020).


  • Coupling non-denaturing chromatography to mass spectrometry for the characterization of monoclonal antibodies and related products.: The study showcases the integration of non-denaturing chromatography with mass spectrometry, utilizing Ion exchanger III to maintain protein integrity and achieve high-resolution characterization of monoclonal antibodies (Farsang et al., 2020).



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron microscopy study of CVT grown Fe-sulphides.
Zavasnik J and Recnik A.
Journal of Crystal Growth, 367, 18-23 (2013)
Eagleson M.
Concise Encyclopedia Chemistry, 124-124 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 93-93 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 199-199 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service