Skip to Content
Merck
All Photos(2)

Documents

200085

Sigma-Aldrich

Mercury(II) bromide

ACS reagent

Synonym(s):

Mercuric bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgBr2
CAS Number:
Molecular Weight:
360.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

vapor pressure

1 mmHg ( 136.5 °C)

form

powder

reaction suitability

reagent type: catalyst
core: mercury

impurities

≤0.05% insol. CH3OH

reduction residue

≤0.02%

color

white to very faintly yellow

bp

322 °C (lit.)

mp

236 °C (lit.)

anion traces

chloride (Cl-): ≤0.25%

SMILES string

Br[Hg]Br

InChI

1S/2BrH.Hg/h2*1H;/q;;+2/p-2

InChI key

NGYIMTKLQULBOO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mercury(II) bromide (HgBr2) affords 1:1 adducts on reaction with N,N,N′,N′- tetramethyl-o-phenylenediamine. Single-crystal X-ray diffraction studies of these adducts have been reported. It forms complexes by reacting with 3,12-dimethylbenzo[a]quinoxalino[2,3-c]phenazine ligand.

Application

Mercury(II) bromide (HgBr2) may be used in the preparation of mononuclear [Hg(L1)Br2] and 1D polymer [Hg2(L2)Br4]n [L1 = (N,N-diethyl,N′-(pyridin-2-yl)formylidene)ethane-1,2-diamine and L2 = (N,N-diethyl,N′-(pyridin-2-yl)benzylidene)ethane-1,2-diamine]. It may also be used as a promoter in the glycoslyation reaction of alcohols.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Quinoxalinophenazine Derivatives and Reaction of 3, 12-Dimethylbenzo [a] quinoxalino [2, 3-c] phenazine with Mercury (II) Bromide: Spectral and Structural Characterization.
Marjani AP, et al.
Chinese Journal of Structural Chemistry / Jie Gou Hua Xue, 33(10), 1460-1466 (2014)
Crystal Structures of the 1: 1 Adducts of N, N, N', N'-Tetramethyl-o-phenylenediamine with Zinc (II) Bromide and Mercury (II) Bromide and Iodide.
Hughes CM, et al.
Australian Journal of Chemistry, 38(10), 1521-1527 (1985)
Syntheses, Structures and Properties of Two Luminous Mercury (II) Bromides Containing Tridentate N-Donor Schiff Bases: Control of Coordination Number and Nuclearity by Varying Ligand Matrices.
Satapathi S, et al.
Journal of Chemical Crystallography, 42(10), 1060-1066 (2012)
Practical synthesis of the 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-?-d-glucosides of Fmoc-serine and Fmoc-threonine and their benzyl esters.
Carvalho I, et al.
Carbohydrate Research, 338(10), 1039-1043 (2003)
S Engst et al.
Biochemistry, 37(33), 11496-11507 (1998-08-26)
Conditions are described under which the nonphysiological substrate mercuric bromide (HgBr2) is rapidly turned over, both by the wild type (CCCC) and by an active site double mutant (CCAA) of mercuric reductase in which the C-terminal cysteines 557' and 558'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service