Skip to Content
Merck
All Photos(1)

Key Documents

13571

Sigma-Aldrich

Sodium sulfate decahydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.0% (calc. to the dried substance)

Synonym(s):

Glauber’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4 · 10H2O
CAS Number:
Molecular Weight:
322.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
crystals

grade

puriss.

Assay

≥99.0% (calc. to the dried substance)

form

crystals

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

acidity or alkalinity, complies
residual solvents, complies
≤10 mg/kg heavy metals (as Pb)
≤200 mg/kg free alkali (as NaOH)
≤300 mg/kg free acid (as H2SO4)

loss

53-57% loss on drying, 130 °C

density

1.46 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Fe: ≤5 mg/kg
Mg: ≤100 mg/kg
Zn: ≤20 mg/kg

suitability

complies for appearance of solution
complies for identity

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

RSIJVJUOQBWMIM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structures of sodium sulfate decahydrate (Glauber's salt) and sodium tetraborate decahydrate (borax). Redetermination by neutron diffraction.
Levy HA and Lisensky GC.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(12), 3502-3510 (1978)
Thermodynamics of crystallization of sodium sulfate decahydrate in H2O-NaCl-Na2SO4: application to Na2SO4?10H2O-based latent heat storage materials.
Marliacy P, et al.
Thermochimica Acta, 344(1), 85-94 (2000)
C Poelaert et al.
Journal of animal science, 90 Suppl 4, 387-389 (2013-02-13)
Over the past decade, several in vitro methods have been developed to study intestinal fermentation in pigs and its influence on health. In these methods, samples are fermented by a bacterial inoculum diluted in a mineral buffer solution. Generally, a
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the
Jia-ying Zhu et al.
Zhongguo wei zhong bing ji jiu yi xue = Chinese critical care medicine = Zhongguo weizhongbing jijiuyixue, 24(6), 346-348 (2012-10-02)
To investigate the effect of nasal tube feeding of rhubarb solution and umbilical compress of Glauber's salt for purgation in the treatment of patients with severe acute organophosphorus pesticide poisoning (AOPP). A retrospective study was conducted. Eighty patients with severe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service