Skip to Content
Merck
All Photos(1)

Key Documents

12038

Sigma-Aldrich

Calcium hydroxide

puriss., meets analytical specification of Ph. Eur., USP, FCC, 95-100.5% (complexometric)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:
NACRES:
NA.21
form:
powder
Pricing and availability is not currently available.

grade

puriss.

Assay

95-100.5% (complexometric)

form

powder

quality

meets analytical specification of Ph. Eur., USP, FCC

impurities

≤0.2% insoluble in hydrochloric acid
≤2.5% subst. not precip. by ammonium oxalate (as SO4)

pH

12.4-12.6 (20 °C)

density

2.24 g/mL at 25 °C (lit.)

anion traces

fluoride (F-): ≤50 mg/kg

cation traces

As: ≤3 mg/kg
Cu: ≤10 mg/kg
Fe: ≤1000 mg/kg
Pb: ≤2 mg/kg
Zn: ≤10 mg/kg

SMILES string

O[Ca]O

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F H van de Sande et al.
Journal of dental research, 92(7 Suppl), 78S-83S (2013-05-22)
This practice-based retrospective study evaluated the survival of resin composite restorations in posterior teeth, focusing on the influence of potential patient risk factors. In total, 306 posterior composite restorations placed in 44 adult patients were investigated after 10 to 18
Lidan Ye et al.
Bioresource technology, 132, 38-44 (2013-02-13)
Cost-effective production of optically pure lactic acid from lignocellulose sugars is commercially attractive but challenging. Bacillus coagulans C106 was isolated from environment and used to produce l-lactic acid from xylose at 50°C and pH 6.0 in mineral salts medium containing
C Sathorn et al.
International endodontic journal, 40(1), 2-10 (2007-01-11)
To determine to what extent does calcium hydroxide intracanal medication eliminate bacteria from human root canals, compared with the same canals before medication, as measured by the number of positive cultures, in patients undergoing root canal treatment for apical periodontitis
Sanaa Chala et al.
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics, 112(4), e36-e42 (2011-07-23)
The aim of this study was to conduct a quantitative systematic review, including published data, comparing the efficacy of mineral trioxide aggregate and calcium hydroxide as material used for the endodontic management of immature teeth. Relevant studies published through November
G H Yassen et al.
International endodontic journal, 46(2), 112-118 (2012-09-14)
The aim of this review was to identify and analyse all studies related to the effect of nonsetting calcium hydroxide [Ca(OH)(2)] on root fracture and various mechanical properties of radicular dentine. A PubMed search was conducted using the keywords 'calcium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service