Skip to Content
Merck
All Photos(1)

Key Documents

11520

Sigma-Aldrich

Lead(II) nitrate

puriss., ≥99%

Synonym(s):

Lead dinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

puriss.

Assay

≥99%

form

solid

reaction suitability

reagent type: catalyst
core: lead

mp

470 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

SMILES string

[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujiao Zhu et al.
Fish & shellfish immunology, 34(2), 617-622 (2013-01-01)
Glucose regulated protein (GRP) located in endoplasmic reticulum (ER) was a member of heat shock protein (Hsp) family. The protective mechanism adapted to ER stimuli was closely related to GRP. GRP78, known as BiP, was one of central regulator responded
Kiyomitsu Nemoto et al.
The Journal of toxicological sciences, 38(1), 127-129 (2013-01-30)
We previously demonstrated the super-induced expression of the Grin2c gene encoding the N-methyl-D-aspartate receptor 2C subunit during the development of liver enlargement with hepatocellular hypertrophy induced by phenobarbital, clofibrate, or piperonyl butoxide. In the present study, we assessed whether or
Xuemei Li et al.
Journal of hazardous materials, 213-214, 55-61 (2012-02-24)
An endophytic fungus was tested in rice (Oryza sativa L.) exposed to four levels of lead (Pb) stress (0, 50, 100 and 200 μM) to assess effects on plant growth, photosynthesis and antioxidant enzyme activity. Under Pb stress conditions, endophyte-infected
Trivedi Mrugesh et al.
Acta poloniae pharmaceutica, 68(5), 653-656 (2011-09-21)
The present study was an attempt to investigate toxic effects of lead on red blood corpuscle (RBC) under in vitro conditions. Venous blood samples from adult, healthy, well-nourished male donors falling in age group of 25-30 years were collected in
Qiang Wang et al.
Brain research, 1455, 19-27 (2012-04-17)
Lead (Pb) can target the vascular system for both acute injury and disease promotion. Cellular iron (Fe) disruption may be implicated in Pb vascular toxicity. To investigate the potential involvement of iron response element 1 (IRP1) protein in the vascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service