Skip to Content
Merck
All Photos(1)

Documents

Z804452

UV Photometric Accuracy Standards

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-2003)

technique(s)

UV/Vis spectroscopy: suitable

application(s)

food and beverages
petroleum
pharmaceutical
plastics

format

neat

General description

CRM for UV/Visible Spectrophotometers.

These UV Photometric Accuracy Standards are design to qualify Photometric Accuracy of UV Spectrophotometers to the limits defined in the EP.

Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.

All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.

Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.

Application

UV Photometric Accuracy Standards for use in the qualification of UV spectrophotometers.

Features and Benefits

  • Product is of certified reference material grade
  • Includes two ampoules of standards and an ampoule with a blank
  • CoA has the instructions on how to qualify the instrument

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service