Z803367
Hellma® liquid calibration standards
UV40, for testing the photometric accuracy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Hellma 667-UV40
technique(s)
UV/Vis spectroscopy: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to photometric accuracy.
Consisting of :
40 mg Potassium dichromate in perchloric acid (667-UV40, wavelength 235; 257; 313; 350 nm)
Consisting of :
40 mg Potassium dichromate in perchloric acid (667-UV40, wavelength 235; 257; 313; 350 nm)
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service