Skip to Content
Merck
All Photos(1)

Documents

Y0001462

Acetylsulfadiazine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4O3S
CAS Number:
Molecular Weight:
292.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfadiazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C12H12N4O3S/c1-9(17)15-10-3-5-11(6-4-10)20(18,19)16-12-13-7-2-8-14-12/h2-8H,1H3,(H,15,17)(H,13,14,16)

InChI key

NJIZUWGMNCUKGU-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetylsulfadiazine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Westerlund et al.
Journal of pharmaceutical sciences, 71(10), 1142-1145 (1982-10-01)
Sulfadiazine and N4-acetylsulfadiazine were determined in biological fluids by the direct injection (plasma after protein precipitation and urine after dilution 100 times) of 20 microliters on a silica gel column. The mobile phase was an aqueous citrate buffer (pH 4.0)
The case: 69 year old man with sand in the urine. N-acetylsulfadiazine crystals.
A K Schuler et al.
Kidney international, 72(6), 769-770 (2007-09-07)
Sulfadiazine/N4-acetylsulfadiazine crystalluria in a patient with the acquired immune deficiency syndrome (AIDS)
J L Potter et al.
Clinica chimica acta; international journal of clinical chemistry, 230(2), 221-224 (1994-10-31)
T Bergan et al.
Chemotherapy, 32(6), 478-485 (1986-01-01)
The study was aimed at analyzing the pharmacokinetics of sulphadiazine (SDZ) and trimethoprim (TMP) in geriatric patients and was carried out in two stages. The first stage compared 11 geriatric patients (68-89 years old) and 9 healthy volunteers (21-34 years
Turner's syndrome with anorexia nervosa.
T Ohzeki et al.
The American journal of medicine, 84(4), 792-793 (1988-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service