Skip to Content
Merck
All Photos(1)

Key Documents

Y0001458

Piretanide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Piretanide, 4-Phenoxy-3-(1-pyrrolidinyl)-5-sulfamoylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O5S
CAS Number:
Molecular Weight:
362.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

piretanide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1c(c(cc(c1)C(=O)O)N3CCCC3)Oc2ccccc2

InChI

1S/C17H18N2O5S/c18-25(22,23)15-11-12(17(20)21)10-14(19-8-4-5-9-19)16(15)24-13-6-2-1-3-7-13/h1-3,6-7,10-11H,4-5,8-9H2,(H,20,21)(H2,18,22,23)

InChI key

UJEWTUDSLQGTOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piretanide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Takamura et al.
Pharmaceutical research, 13(7), 1015-1019 (1996-07-01)
The purpose of this study was to investigate the binding mechanism of loop diuretics with HSA and to characterize the binding site on HSA. Quantitative analysis of potential interaction between ligands bound to HSA was performed by equilibrium dialysis and
V Homuth et al.
The American journal of cardiology, 72(9), 666-671 (1993-09-15)
To test the dose responses of piretanide, ramipril, and their combination in patients with essential hypertension, a prospective, randomized, double-blind, placebo-controlled trial was conducted in 480 patients. Twelve separate groups were studied: placebo, piretanide 3 mg, piretanide 6 mg, ramipril
G Ruf et al.
European journal of clinical pharmacology, 46(6), 545-550 (1994-01-01)
The pharmacokinetics and pharmacodynamics of single oral doses of 5 mg ramipril and 6 mg piretanide administered separately and in combination were determined in a single blind, randomised, 3-period cross-over study in 24 healthy male volunteers. The peak plasma concentrations
P Hannaert et al.
Naunyn-Schmiedeberg's archives of pharmacology, 365(3), 193-199 (2002-03-08)
It is generally assumed that bumetanide possesses some selectivity for the renal Na-K-Cl cotransporter NKCC2, although the results are scarce in the literature and comparisons were done with extra-renal NKCC1 at its basal, almost silent state. Here we investigated NKCC2/NKCC1
F Pourageaud et al.
British journal of pharmacology, 131(6), 1211-1219 (2000-11-18)
1. Relaxing effect of loop diuretics, piretanide and furosemide in comparison with acetylcholine (ACh) was investigated in guinea-pig isolated mesenteric resistance arteries. 2. Concentration-response curves to ACh (0.001 - 10 microM) and diuretics (0.0001 - 1 microM) were constructed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service