Skip to Content
Merck
All Photos(1)

Key Documents

Y0001373

Nicergoline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrN3O3
CAS Number:
Molecular Weight:
484.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

nicergoline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12Cc3cn(C)c4cccc(c34)[C@]1(C[C@@H](COC(=O)c5cncc(Br)c5)CN2C)OC

InChI

1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1

InChI key

YSEXMKHXIOCEJA-FVFQAYNVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicergoline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Treatment of impaired cognition with nootropic drugs: nicergoline versus the state of the art.
R G Fariello
Functional neurology, 12(3-4), 221-225 (1997-05-01)
M Fioravanti et al.
The Cochrane database of systematic reviews, (4)(4), CD003159-CD003159 (2001-11-01)
Nicergoline is an ergot derivative currently in use in over fifty countries for more than three decades, for the treatment of cognitive, affective, and behavioral disorders of older people. It was initially considered as a vasoactive drug and mainly prescribed
Bengt Winblad et al.
Clinical drug investigation, 28(9), 533-552 (2008-08-01)
The ergot alkaloid derivative nicergoline became clinically available about 35 years ago in the 1970s. Nicergoline has a broad spectrum of action: (i) as an alpha(1)-adrenoceptor antagonist, it induces vasodilation and increases arterial blood flow; (ii) it enhances cholinergic and
[Nicergoline, ibudilast, ifenprodil tartrate].
Takako Arii et al.
Nihon rinsho. Japanese journal of clinical medicine, 64 Suppl 7, 602-605 (2007-04-28)
A Moretti et al.
Arzneimittel-Forschung, 29(8a), 1223-1227 (1979-01-01)
10-Methoxy-1,6-dimethyl-ergoline-8 beta-methanol-(5-bromonicotinate) (nicergoline, Sermion) shows a strong alpha-blocking activity both in vitro and in vivo. Various studies (dog, cat, rabbit, rat, mouse, guinea-pig) show that nicergoline affects only slightly blood pressure and heart rate and increases the blood flow in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service