Skip to Content
Merck
All Photos(1)

Key Documents

Y0001058

Dextropropoxyphene hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO2 · HCl
CAS Number:
Molecular Weight:
375.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dextropropoxyphene

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].N(C[C@H]([C@@](OC(=O)CC)(Cc2ccccc2)c1ccccc1)C)(C)C.[H+]

InChI

1S/C22H29NO2.ClH/c1-5-21(24)25-22(18(2)17-23(3)4,20-14-10-7-11-15-20)16-19-12-8-6-9-13-19;/h6-15,18H,5,16-17H2,1-4H3;1H/t18-,22+;/m1./s1

InChI key

QMQBBUPJKANITL-MYXGOWFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextropropoxyphene hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nick A Buckley et al.
The Medical journal of Australia, 199(4), 257-260 (2013-08-30)
• The Therapeutic Goods Administration determined in November 2011 that dextropropoxyphene should be removed from the Australian Register of Therapeutic Goods. This is consistent with this drug's removal from the market in many other developed countries. • However, dextropropoxyphene is
P Bertin et al.
The journal of nutrition, health & aging, 17(8), 681-686 (2013-10-08)
The main objective of the S.AGES (Elderly Subjects) cohort study is to describe the current therapeutic strategy for chronic pain in non-institutionalised elderly patients in France. In this prospective cohort study, non-institutionalised patients aged 65 years and over with chronic
Keith Hawton et al.
PLoS medicine, 9(5), e1001213-e1001213 (2012-05-17)
The analgesic co-proxamol (paracetamol/dextropropoxyphene combination) has been widely involved in fatal poisoning. Concerns about its safety/effectiveness profile and widespread use for suicidal poisoning prompted its withdrawal in the UK in 2005, with partial withdrawal between 2005 and 2007, and full
Surendra Kumar Mattoo et al.
General hospital psychiatry, 35(1), 100-101 (2012-12-04)
As an infrequent symptom diplopia has been reported with opiate withdrawal, especially heroin, but not dextropropoxyphene. We report incomitant esotropia and diplopia in a case with dextropropoxyphene withdrawal that resolved completely with the resolution of the opiate withdrawal syndrome.
Stephen Winbery et al.
Southern medical journal, 104(7), 533-539 (2011-09-03)
Medicare Part D data from the Quality Improvement Organization's 9th Statement of Work drug safety indicator project under the direction of the Centers for Medicare & Medicaid Services define the potentially inappropriate medications (PIMs) list for Tennessee. These data reveal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service