Skip to Content
Merck
All Photos(1)

Documents

Y0000837

Triamterene

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-pteridinetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N7
CAS Number:
Molecular Weight:
253.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triamterene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1nc(N)c2nc(-c3ccccc3)c(N)nc2n1

InChI

1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)

InChI key

FNYLWPVRPXGIIP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamterene EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Pharmacology of potassium-sparing diuretics: amiloride and triamterene].
G Rorive et al.
Coeur et medecine interne, 17(2), 207-215 (1978-04-01)
H Priewer et al.
Die Pharmazie, 52(3), 179-181 (1997-03-01)
This work represents a summary of studies carried out with potassium and magnesium retaining triamterene derivatives. The synthesis, analytics and physico-chemical characteristics of those substances are described. Furthermore, the relationships between the chemical structure and electrolyte excretion, pharmacokinetics, metabolism and
D A Sica et al.
Nephron, 51(4), 454-461 (1989-01-01)
Triamterene (TA) is a mild 'potassium-sparing' diuretic usually employed in combination with other more potent diuretics in the treatment of hypertension. TA pharmacokinetics and pharmacodynamics in normal volunteers, elderly subjects and in patients with renal and hepatic dysfunction are reviewed.
C Lucas et al.
Journal d'urologie, 88(1), 37-42 (1982-01-01)
Various studies have demonstrated a relationship between nephrolithiasis and the ingestion of certain drugs. We are particularly interested in the effects of triamterene. Five published case studies on patients of both sexes between 43 and 60 years of age have
A Haris et al.
Orvosi hetilap, 137(35), 1907-1914 (1996-09-01)
The group of drugs, so-called "potassium sparing diuretics" represent an important part of our modern therapeutic arsenal. Their "weak diuretic" properties are especially beneficial in cirrhotic patients with ascites, when highly effective loop diuretics may be hazardous. Potassium sparing diuretics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service