Skip to Content
Merck
All Photos(1)

Documents

Y0000803

Ethambutol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ethambutol dihydrochloride, 2,2′-(1,2-Ethanediyldiimino)bis-1-butanol dihydrochloride, Emb

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2O2 · 2HCl
CAS Number:
Molecular Weight:
277.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ethambutol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC[C@@H](CO)NCCN[C@H](CO)CC.Cl.Cl

InChI

1S/C10H24N2O2.2ClH/c1-3-9(7-13)11-5-6-12-10(4-2)8-14;;/h9-14H,3-8H2,1-2H3;2*1H/t9-,10-;;/m0../s1

InChI key

AUAHHJJRFHRVPV-BZDVOYDHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethambutol is an antimycobacterial used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance. It has been used to study multidrug-resistant tuberculosis (MDR-TB) in highest-incidence-rate areas in China.

Biochem/physiol Actions

Ethambutol is bacteriostatic against actively growing TB bacilli. Mycolic acids attach to the 5′-hydroxyl groups of D-arabinose residues of arabinogalactan and form mycolyl-arabinogalactan-peptidoglycan complex in the cell wall. Ethambutol disrupts arabinogalactan synthesis by inhibiting arabinosyltransferases, specifically those in Mycobacterium, interfering with the biosynthesis of arabinogalactans found in the cell wall of mycobacteria and leading to increased cell wall permeability.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wajeeh R Bakhsh et al.
Journal of surgical orthopaedic advances, 23(3), 162-165 (2014-08-26)
Mycobacterium kansasii is an acid-fast bacillus most commonly associated with pulmonary pathology. Infection of the spine is exceedingly rare, with just three reported cases, two of which were in human immunodeficiency virus and acquired immunodeficiency syndrome patients. This case report
Mohammed Shafik El-Ridy et al.
Drug delivery, 22(1), 21-36 (2013-12-24)
Tuberculosis (TB) is a worldwide health concern. In 2011, about 8.7 million new cases developed TB and 1.4 million people died from it. Enhancement of ethambutol hydrochloride activity and safety in treatment of TB through niosomal encapsulation. Niosomes were prepared
N Guo et al.
Letters in applied microbiology, 58(4), 344-349 (2013-12-18)
Previous studies have shown that isoimperatorin (IO), a furanocoumarin isolated from several medicinal plants, has antimycobacterial activity against Mycobacterium tuberculosis strain H37Rv (ATCC 27294). This study demonstrated that IO has antimycobacterial activity against 2 drug-sensitive and 6 drug-resistant isolates, with
Joshua R Francis et al.
The Medical journal of Australia, 200(6), 328-332 (2014-04-08)
OBJECTIVE To describe the epidemiology, clinical features, health care resource use, treatment and outcomes of multidrug-resistant tuberculosis (MDR-TB) cases diagnosed in Western Australia, compared with matched controls with drug-susceptible TB. Retrospective case-control study of all MDR-TB cases notified in WA
Partha Pal et al.
The American journal of tropical medicine and hygiene, 90(5), 788-789 (2014-05-09)
A 60-year-old woman presented with a painful lump of 4 months duration in her right breast. She denied any history of breast trauma or family history of breast cancer. Physical examination detected a firm, tender mass in the upper inner

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service