Skip to Content
Merck
All Photos(1)

Documents

Y0000570

Clobetasol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clobetasol propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl

InChI

1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1

InChI key

CBGUOGMQLZIXBE-XGQKBEPLSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon H Kircik et al.
Journal of drugs in dermatology : JDD, 12(3), 328-334 (2013-04-03)
Chronic hand dermatitis may have a significant detrimental effect on daily home-related and work-related activities, and quality of life (QOL). Clobetasol propionate foam, 0.05%, is indicated for the treatment of inflammatory and pruritic manifestations of corticosteroid-responsive dermatoses in patients aged
S R Wiegell et al.
The British journal of dermatology, 171(6), 1487-1492 (2014-07-26)
Photodynamic therapy (PDT) is an effective and established treatment for actinic keratoses (AK) and nonmelanoma skin cancer. The main side-effects of PDT are post-treatment erythema and oedema, and pain during illumination. Severe erythema after PDT enhances the down time associated
Sidharth Sonthalia et al.
International journal of dermatology, 51(11), 1371-1378 (2012-10-17)
Oral lichen planus (OLP) is a common disease of the oral mucosa with worldwide distribution and overall prevalence of 0.5-2.2%. Its etiology remains unclear, although the role of autoimmunity is supported by its association with other autoimmune diseases and the
Mark G Lebwohl et al.
Journal of the American Academy of Dermatology, 69(3), 385-392 (2013-05-07)
Few clinical trials have evaluated the combination of topical corticosteroids plus systemic therapies for psoriasis. We sought to evaluate efficacy and safety of etanercept plus topical clobetasol propionate (CP) foam versus etanercept monotherapy for treatment of moderate to severe plaque
Sem Decani et al.
The Journal of dermatological treatment, 25(6), 495-500 (2012-12-06)
Topical corticosteroids are considered first-line therapy in patients with chronic inflammatory oral mucosal diseases; among them, clobetasol propionate is one of the most widely used in oral medicine. Under physiological conditions, the transmucosal application is characterized by a significantly greater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service