Skip to Content
Merck
All Photos(1)

Key Documents

Y0000537

Emedastine difumarate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34N4O9
CAS Number:
Molecular Weight:
534.56
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

emedastine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1(CCN(CCC1)c2[n](c3c(n2)cccc3)CCOCC)C.OC(=O)\C=C\C(=O)O.OC(=O)\C=C\C(=O)O

InChI

1S/C17H26N4O.2C4H4O4/c1-3-22-14-13-21-16-8-5-4-7-15(16)18-17(21)20-10-6-9-19(2)11-12-20;2*5-3(6)1-2-4(7)8/h4-5,7-8H,3,6,9-14H2,1-2H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+

InChI key

FWLKKPKZQYVAFR-LVEZLNDCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Emedastine difumarate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annik Pons-Guiraud et al.
European journal of dermatology : EJD, 16(6), 649-654 (2007-01-19)
Emedastine difumarate (2 mg b.i.d.) was compared to loratadine (10 mg o.d.) in a randomized, double-blind, multicentre trial for 4 weeks in 192 patients with idiopathic chronic urticaria. After one week of treatment significant differences were recorded: body skin involvement
Mario E Corrado et al.
Arzneimittel-Forschung, 54(10), 660-665 (2004-11-24)
Emedastine is a new H1-receptor antagonist endowed with potent and selective antihistamine activity. The aim of this study was to evaluate the therapeutic efficacy and tolerability of emedastine difumarate (CAS 87233-62-3) in Caucasian patients in the treatment of seasonal allergic
Shoichi Harada et al.
Journal of pharmaceutical sciences, 94(1), 17-24 (2005-03-12)
Antihistamine effects of emedastine applied topically with three vehicles varying in their polarities were investigated in rats. The pharmacological effect of emedastine differed greatly depending on its concentration, treatment time, and vehicle. The antihistamine effect reached a plateau after approximately
László Endre
Orvosi hetilap, 148(6), 251-254 (2007-03-09)
In the treatment of any disease, it is advantageous if the concentration of the administered drug is highest in the target organ. It follows that, if possible, diseases of the conjunctiva should primarily be treated with eye drops. The author
Yuan Tian et al.
Journal of chromatographic science, 45(3), 158-164 (2007-04-28)
A selective and sensitive method employing high-performance liquid chromatography (HPLC)-electrospray ionization (ESI)-mass spectrometry is developed and validated for the determination of emedastine difumarate in human plasma. With naphazoline hydrochloride as the internal standard, emedastine difumarate is extracted from plasma with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service