Skip to Content
Merck
All Photos(1)

Key Documents

Y0000462

Torasemide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Torsemide, Demadex, N-[[(1-Methylethyl)amino]carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N4O3S
CAS Number:
Molecular Weight:
348.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

torasemide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2

InChI

1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-8-7-14(15)19-13-6-4-5-12(3)9-13/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)

InChI key

NGBFQHCMQULJNZ-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Torasemide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Torsemide is a loop diuretic and inhibitor of the Na+/K+/2Cl- carrier system.
Torsemide is a loop diuretic of the pyridine-sulfonylurea class with antialdosteronergic properties and inhibitor of the Na+/K+/2Cl- carrier system.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark A Oyama et al.
Journal of veterinary cardiology : the official journal of the European Society of Veterinary Cardiology, 13(4), 287-292 (2011-10-28)
Diuretics are a mainstay of therapy in dogs with heart failure. In dogs with advanced heart failure, moderate to high doses of loop diuretics such as furosemide are used with diminishing effects as profound activation of neuroendocrine systems promote signs
Zarko Jovic et al.
Journal of chromatographic science, 50(4), 324-334 (2012-03-13)
Complete evaluation of chromatographic behavior and establishment of optimal experimental conditions for determination of torasemide and its four impurities are determined by experimental design. Fractional factorial and 3(n) full factorial design were employed for efficient and rapid optimization of liquid
Hitesh B Patel et al.
Journal of AOAC International, 94(1), 143-149 (2011-03-12)
A simple RP-ultra-performance LC method was developed and validated for determination of impurities related to torsemide tablets. The rapid method provided adequate separation of all known related impurities and degradation products. Separation was achieved on a Zorbax SB-C18 column (50
Sanjay Gupta et al.
European journal of heart failure, 12(7), 746-752 (2010-06-08)
Diuretics, when used to treat congestion in patients with chronic heart failure, improve symptoms and, perhaps, prognosis but little information is available to guide their use in patients with left ventricular systolic dysfunction (LVSD) who are not congested. Chronic diuretic
Carmen Acuña
Drugs of today (Barcelona, Spain : 1998), 46 Suppl E, 1-15 (2010-10-12)
Chronic heart failure (CHF) is the cause of significant morbimortality all over the world and its incidence and prevalence are increasing. Fluid retention and volume overload are responsible of large part of morbility related to heart failure. There are a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service