Skip to Content
Merck
All Photos(1)

Documents

Y0000439

Didanosine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′,3′-Dideoxyinosine, ddI, ddIno

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O3
CAS Number:
Molecular Weight:
236.23
Beilstein:
3619529
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

didanosine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H]1CC[C@@H](O1)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H12N4O3/c15-3-6-1-2-7(17-6)14-5-13-8-9(14)11-4-12-10(8)16/h4-7,15H,1-3H2,(H,11,12,16)/t6-,7+/m0/s1

InChI key

BXZVVICBKDXVGW-NKWVEPMBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Didanosine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tenofovir and didanosine: a dangerous liaison.
Laura Waters et al.
The AIDS reader, 15(8), 403-406 (2005-08-23)
J J McGowan et al.
Reviews of infectious diseases, 12 Suppl 5, S513-S520 (1990-07-01)
AIDS has remained a significant and worsening medical problem since its first description as a new clinical entity in 1981. In the past 6 years, substantial progress has been made in the chemotherapy for this disease; such progress is likely
William R Birmingham et al.
Nature chemical biology, 10(5), 392-399 (2014-03-25)
Concatenation of engineered biocatalysts into multistep pathways markedly increases their utility, but the development of generalizable assembly methods remains a major challenge. Herein we evaluate 'bioretrosynthesis', which is an application of the retrograde evolution hypothesis, for biosynthetic pathway construction. To
Fanconi syndrome and nephrogenic diabetes insipidus associated with didanosine therapy in HIV infection: a case report and literature review.
Géraldine D'Ythurbide et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 22(12), 3656-3659 (2007-10-02)
Marco Bongiovanni et al.
Current medicinal chemistry, 13(23), 2789-2793 (2006-11-01)
Nucleoside reverse transcriptase inhibitors (NRTI) are essential components of highly active antiretroviral treatment (HAART). Although several combinations can be used as NRTI backbones, not all are associated with good virological and/or immunological results. In particular, some NRTI combinations should be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service