Skip to Content
Merck
All Photos(1)

Key Documents

Y0000384

All-rac-alpha-tocopheryl acetate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

DL-α-Tocopherol acetate, Vitamin E acetate, all-rac-α-Tocopheryl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

30 MG
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onApril 18, 2025


Request a Bulk Order

Select a Size

Change View
30 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
Beilstein:
97512
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onApril 18, 2025


Request a Bulk Order

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

tocopherol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

density

0.96 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
All-rac-alpha-tocopheryl acetate, the ester of all-rac-alpha-tocopherol, is a commercial form of vitamin E. It is responsible for the protection of phenol groups of the parent tocopherol against oxidation.[1][2]

Application

All-rac-alpha-tocopheryl acetate for peak identification may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.[3]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

all-rac-?-Tocopheryl acetate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 129(7), 3802-3804 (2011)
All-rac-alpha-tocopherol acetate is a better vitamin E source than all-rac-alpha-tocopherol succinate for broilers
Jensen S, et al.
The Journal of Nutrition, 129(7), 1355-1360 (1999)
Succinic Acids?Advances in Research and Application: 2013 Edition: ScholarlyBrief
Acton QA
Succinic Acids?Advances in Research and Application (2013)
Po-Hsien Huang et al.
Science signaling, 6(267), ra19-ra19 (2013-03-21)
Vitamin E is a fat-soluble vitamin with antioxidant properties. Tocopherols are the predominant form of vitamin E found in the diet and in supplements and have garnered interest for their potential cancer therapeutic and preventive effects, such as the dephosphorylation
Adam Prokopowicz et al.
Occupational and environmental medicine, 70(6), 365-371 (2013-02-05)
Changes in enzymatic antioxidant activity are frequently observed in workers occupationally exposed to lead. Few studies have investigated the influence of lead on the non-enzymatic antioxidant system. The aim of our study was to assess the influence of occupational exposure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service