Skip to Content
Merck
All Photos(1)

Key Documents

Y0000366

Isradipine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(4-Benzofurazanyl)-1,-4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid methyl 1-methylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3O5
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

isradipine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2cccc3nonc23)C(=O)OC(C)C

InChI

1S/C19H21N3O5/c1-9(2)26-19(24)15-11(4)20-10(3)14(18(23)25-5)16(15)12-7-6-8-13-17(12)22-27-21-13/h6-9,16,20H,1-5H3

InChI key

HMJIYCCIJYRONP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isradipine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

L-type calcium channel blocker (also referred to as dihydropyridine-type calcium channel blocker); antihypertensive.
L-type calcium channel blocker; antihypertensive.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Marty
Acta anaesthesiologica Scandinavica. Supplementum, 99, 53-55 (1993-01-01)
The traditional therapy of perioperative hypertension is now being challenged by the availability of new agents. Vasodilator therapy should be the first-line choice, although dihydropyridine calcium antagonists, particularly isradipine, appear to be suitable in this setting because of their pharmacological
R P Hof et al.
American journal of hypertension, 4(2 Pt 2), 107S-113S (1991-02-01)
In the context of atherosclerosis, calcium antagonists should be evaluated from two points of view: their hemodynamic action on the atherosclerotic cardiovascular system, and their preventive action on atherosclerosis. Contraction of atherosclerotic vessels in response to vasoconstrictors, especially serotonin and
W H Frishman
Journal of clinical pharmacology, 34(12), 1164-1172 (1994-12-01)
In the treatment of hypertension in renally impaired patients, normalization of blood pressure alone may not be sufficient to prevent significant morbidity to the kidneys. Treatment must reduce pressure in the renal vasculature, otherwise glomerular filtration rate and renal plasma
D Panuccio et al.
La Clinica terapeutica, 146(11), 729-736 (1995-11-01)
The aim of the present study was to evaluate efficacy on blood pressure values and metabolic tolerability of Isradipine, which was given to 15 mild hypertensive non diabetic subjects (average age 63.8 +/- 10.9 years), at the dosage of 5
D Blecker
Angiology, 45(12), 997-1008 (1994-12-01)
Treatment of hypertension in the complicated patient requires a specialized approach focusing on the treatment of the blood pressure and concurrent medical problems. Therapy must be chosen in consideration of the patient's age, multiple illnesses, declining organ function, and possible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service