Skip to Content
Merck
All Photos(1)

Key Documents

Y0000239

Celiprolol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Celiprolol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33N3O4 · HCl
CAS Number:
Molecular Weight:
415.95
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

celiprolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H

InChI key

VKJHTUVLJYWAEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Celiprolol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

En-Hui Yao et al.
American journal of hypertension, 21(9), 1062-1068 (2008-07-19)
Endothelial progenitor cells (EPCs) derived from bone marrow migrate to areas of endothelial damage and repair them. EPC function is impaired by oxidative stress. We examined the effects of an antioxidative beta1-adrenoceptor blocker on the number and function of EPCs
A Jamard et al.
Journal de gynecologie, obstetrique et biologie de la reproduction, 41(7), 676-678 (2012-04-24)
The Ehlers-Danlos syndrome (EDS) is a rare inheritable disease, characterised by a defect in collagen synthesis. Various types have been described and the type IV or vascular type is the most severe characterised by vascular, gastrointestinal and gynaecologic complications. We
Jiangeng Huang et al.
International journal of pharmaceutics, 356(1-2), 351-353 (2008-02-05)
The aim of this work was to investigate the effects of pluronic F68 block copolymer on the P-gp-mediated transport of celiprolol (CEL) and CYP3A4-mediated formation of midazolam (MDZ) metabolite 1'-hydroxymidazolam. Over a range from 0.03 to 0.3%, pluronic F68 increased
Kim-Thanh Ong et al.
Lancet (London, England), 376(9751), 1476-1484 (2010-09-10)
Vascular Ehlers-Danlos syndrome is a rare severe disease that causes arterial dissections and ruptures that can lead to early death. No preventive treatment has yet been validated. Our aim was to assess the ability of celiprolol, a β(1)-adrenoceptor antagonist with
Jean-Christophe Charniot et al.
Acta cardiologica, 61(6), 661-663 (2007-01-09)
Systemic lupus erythematosus can be idiopathic or drug-induced. Although a number of beta-blockers have been reported to induce a lupus-like syndrome, to the best of our knowledge, no such case has been described following celiprolol therapy. We diagnosed a lupus-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service