Skip to Content
Merck
All Photos(1)

Key Documents

W562125

Sigma-Aldrich

10-Undecen-1-ol

≥98%, stabilized, FG

Synonym(s):

11-Hydroxy-1-undecene, NSC 66157, Undecylenyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)9OH
CAS Number:
Molecular Weight:
170.29
Beilstein:
1753225
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
02.125

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

contains

alpha-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.45 (lit.)

bp

132-133 °C/15 mmHg (lit.)
245-248 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

fragrance allergen

no known allergens

Organoleptic

fresh; green; musty; floral; citrus; waxy

SMILES string

OCCCCCCCCCC=C

InChI

1S/C11H22O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,12H,1,3-11H2

InChI key

GIEMHYCMBGELGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service