Skip to Content
Merck
All Photos(1)

Documents

W430415

Sigma-Aldrich

trans-2-Decenol

natural (US), ≥97%, FG

Synonym(s):

trans-2-Decen-1-ol, (E)-dec-2-en-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH=CHCH2OH
CAS Number:
Molecular Weight:
156.27
FEMA Number:
4304
MDL number:
UNSPSC Code:
12164502
Flavis number:
02.137
NACRES:
NA.21

grade

FG
Kosher
natural (US)

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.451 (lit.)

bp

231-232 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; citrus; herbaceous; waxy

SMILES string

CCCCCCC\C=C\CO

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h8-9,11H,2-7,10H2,1H3/b9-8+

InChI key

QOPYYRPCXHTOQZ-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service