Skip to Content
Merck
All Photos(1)

Key Documents

W417080

Sigma-Aldrich

Methyl N-acetylanthranilate

≥99%, FG

Synonym(s):

Acetyl methyl anthranilate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$87.40
W417080-SAMPLE
$173.00
1 KG
$281.00

$87.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$87.40
W417080-SAMPLE
$173.00
1 KG
$281.00

About This Item

Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
FEMA Number:
4170
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.649
NACRES:
NA.21
grade:
FG
food allergen:
no known allergens

$87.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

FG

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; floral

SMILES string

N(c1c(cccc1)C(=O)OC)C(=O)C

InChI

1S/C10H11NO3/c1-7(12)11-9-6-4-3-5-8(9)10(13)14-2/h3-6H,1-2H3,(H,11,12)

InChI key

UYQKZKVNYKOXHG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service