Skip to Content
Merck
All Photos(1)

Key Documents

T5891

Sigma-Aldrich

True Blue diaceturate salt

Synonym(s):

1,2-Bis(5-amidino-2-benzofuranyl)ethylene diaceturate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N4O2 · 2C4H7NO3
CAS Number:
Molecular Weight:
578.57
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~40%

SMILES string

CC(=O)NCC(O)=O.CC(=O)NCC(O)=O.NC(=N)c1ccc2oc(\C=C\c3cc4cc(ccc4o3)C(N)=N)cc2c1

InChI

1S/C20H16N4O2.2C4H7NO3/c21-19(22)11-1-5-17-13(7-11)9-15(25-17)3-4-16-10-14-8-12(20(23)24)2-6-18(14)26-16;2*1-3(6)5-2-4(7)8/h1-10H,(H3,21,22)(H3,23,24);2*2H2,1H3,(H,5,6)(H,7,8)/b4-3+;;

InChI key

DKAHNYWQHXQOPB-CZEFNJPISA-N

Application

A fluorescent neuronal tracer

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Ando et al.
The Journal of comparative neurology, 528(10), 1775-1802 (2020-01-07)
A significant population of neurons in the vestibular nuclei projects to the cerebellum as mossy fibers (MFs) which are involved in the control and adaptation of posture, eye-head movements, and autonomic function. However, little is known about their axonal projection
Youlai Yu et al.
American journal of translational research, 7(1), 53-65 (2015-03-11)
Functional recovery is often unsatisfactory after lesions in the peripheral nervous system despite the strong potential for regeneration and advances in microsurgical techniques. Axonal regeneration in mixed nerve into inappropriate pathways is a major contributing factor to this failure. In
Kuypers, H.G.J.M. and Huisman, A.M.
Adv. Cellular Neurobiol., 5, 307-307 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service