Skip to Content
Merck
All Photos(1)

Key Documents

STS0019

Sigma-Aldrich

TERGITOL 15-S-3

greener alternative

Non-ionic surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$56.70
500 ML
$84.40

$56.70


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$56.70
500 ML
$84.40

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28

$56.70


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

viscosity

49 cP(25 °C)

transition temp

pour point -46 °C
cloud point insoluble

solubility

chlorinated solvents: soluble (and most organic solvents and oils)

density

0.918 g/mL at 20 °C

HLB

8.0

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service