Skip to Content
Merck
All Photos(1)

Key Documents

SMB01364

Sigma-Aldrich

Leukoaminochrome Hydrochloride

≥95% (HPLC)

Synonym(s):

5,6-Dihydroxyindoline HCl, 5,6-Indolinediol HCl, Indoline-5,6-diol HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2 · xHCl
CAS Number:
Molecular Weight:
151.16 (free base basis)
UNSPSC Code:
12352205
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

storage temp.

2-8°C

SMILES string

OC1=CC2=C(NCC2)C=C1O.Cl

InChI

1S/C8H9NO2.ClH/c10-7-3-5-1-2-9-6(5)4-8(7)11;/h3-4,9-11H,1-2H2;1H

InChI key

IMGPXUZVVPIKOV-UHFFFAOYSA-N

General description

Leukoaminochrome, also referred to as 5,6-Dihydroxyindoline, is a triacetyl derivative within the indoline class of organic compounds. It is a primary metabolite and finds common usage in the cosmetic industry, particularly in hair dye formulations.

Application

Leukoaminochrome can be used in metabolomics research, with a focus on investigating Parkinson′s disease.

Biochem/physiol Actions

Has role in dopamine oxidation and parkinson′s disease

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with a wide variety of chromatographic and spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.
This product is intended for research purposes only, and it is not meant for human consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service