Skip to Content
Merck
All Photos(1)

Key Documents

SMB01340

Sigma-Aldrich

Isoleucyl-Phenylalanine

Synonym(s):

L-Isoleucyl-L-phenylalanine, Ile-Phe, Isoleucylphenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O3
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

Assay

≥95% (HPLC)

Quality Level

form

solid

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](N)C(N[C@@H](CC1=CC=CC=C1)C(O)=O)=O

InChI key

WMDZARSFSMZOQO-DRZSPHRISA-N

General description

Isoleucyl-Phenylalanine is a dipeptide derived from the incomplete breakdown of protein digestion or protein catabolism. It has not yet been identified in human tissues or biofluids and so it is classified as an ′Expected′ metabolite.

Application

Isoleucyl-Phenylalanine can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashmi Sinha et al.
PloS one, 11(3), e0152126-e0152126 (2016-03-26)
Investigation of microbe-metabolite relationships in the gut is needed to understand and potentially reduce colorectal cancer (CRC) risk. Microbiota and metabolomics profiling were performed on lyophilized feces from 42 CRC cases and 89 matched controls. Multivariable logistic regression was used
James J Goedert et al.
Carcinogenesis, 35(9), 2089-2096 (2014-07-20)
Metabolomic analysis of feces may provide insights on colorectal cancer (CRC) if assay performance is satisfactory. In lyophilized feces from 48 CRC cases, 102 matched controls, and 48 masked quality control specimens, 1043 small molecules were detected with a commercial
Dustin G Brown et al.
Cancer & metabolism, 4, 11-11 (2016-06-09)
Colorectal cancers (CRC) are associated with perturbations in cellular amino acids, nucleotides, pentose-phosphate pathway carbohydrates, and glycolytic, gluconeogenic, and tricarboxylic acid intermediates. A non-targeted global metabolome approach was utilized for exploring human CRC, adjacent mucosa, and stool. In this pilot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service