Skip to Content
Merck
All Photos(1)

Key Documents

S5145

Sigma-Aldrich

Silver enhancer solution B

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Silver enhancer solution B,

form

liquid

Quality Level

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

Application

Silver enhancer solution B has been used in the enhancement of metallic nanoparticles[1] and silver staining.[2]

Other Notes

Initiator solution for Silver Enhancer Kit SE-100

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxin detection by Si photosensitive biosensors with a new measurement scheme
Park, et al.
Biosensors And Bioelectronics, 29(1), 219-223 (2011)
DNA microarrays for visual detection of human pathogenic microorganisms based on tyramine signal amplification coupled with gold label silver stain
Qi Hong-Juan, et al.
Analytical and Bioanalytical Chemistry, 398(6), 2745-2750 (2010)
Jeong Moon et al.
Macromolecular bioscience, 19(6), e1800486-e1800486 (2019-04-19)
For the construction of high-performance biosensor, it is important to interface bioreceptors with the sensor surface densely and in the optimal orientation. Herein, a simple surface modification method that can optimally immobilize antibodies onto various kinds of surfaces is reported.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service