Skip to Content
Merck
All Photos(1)

Key Documents

S4251

Sigma-Aldrich

Starch from potato

Powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

potato

Quality Level

form

powder

color

white

pH

5.8-7.8

Looking for similar products? Visit Product Comparison Guide

Application


  • Eco-Friendly, Biodegradable Starch-Based Packaging Materials with Antioxidant Features: Describes the development of potato starch-based packaging that is not only biodegradable but also incorporates antioxidant properties to enhance food preservation (Bajer, 2024).

  • Facile superhydrophobic modification on HPMC film using polydimethylsiloxane and starch granule coatings: Discusses a novel application in modifying surface properties of biodegradable films using starch, thus improving their utility in various industrial and packaging applications (Ye et al., 2024).

  • In-Silico Identification, Characterization and Expression Analysis of Genes Involved in Resistant Starch Biosynthesis in Potato (Solanum tuberosum L.) Varieties: This research provides insights into the genetic basis of starch properties in potatoes, which can lead to the cultivation of varieties with tailored starch characteristics for specific industrial uses (Kaur et al., 2024).

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Haiteng Li et al.
Carbohydrate polymers, 225, 115244-115244 (2019-09-16)
Two models for the digestion of starch in foods are considered: sequential, where two or more reactions follow one another, and parallel, where they occur simultaneously. Each reaction is assumed to be first-order, and to be characterized by a rate
Marcella Brescia et al.
International journal of molecular sciences, 21(19) (2020-10-01)
Duchenne muscular dystrophy (DMD) is the most frequent and severe form of muscular dystrophy. The disease presents with progressive body-wide muscle deterioration and, with recent advances in respiratory care, cardiac involvement is an important cause of morbidity and mortality. DMD
Eiji Yamazaki et al.
Carbohydrate polymers, 94(1), 555-560 (2013-04-03)
Effect of the polysaccharide from leaves of Corchorus olitorius L. (PLC) on the freeze-thaw (FT) stability of corn starch gel was studied. PLC was incorporated into the starch gel at 0.7% and total solid was adjusted to 6.0%. The syneresis
Bruno R Cruz et al.
Carbohydrate polymers, 94(1), 594-602 (2013-04-03)
Starch isolation methods can change their physico-chemical and functional characteristics hindering the establishment of a starch-food functionality relation. A simple high yield and soft isolation method was applied for chestnut (Castanea sativa Mill) starch consisting in steeping and fruit disintegration
Amanda Bortolini Silveira et al.
PLoS genetics, 9(4), e1003437-e1003437 (2013-04-18)
Epigenetic variation, such as heritable changes of DNA methylation, can affect gene expression and thus phenotypes, but examples of natural epimutations are few and little is known about their stability and frequency in nature. Here, we report that the gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service