Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 mL | Check Cart for Availability | $321.00 |
About This Item
grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
7.520 mPa.s(25.00 °C)
application(s)
cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care
format
mixture
General description
1 of 1
This Item | |||
|---|---|---|---|
| description viscosity 7.520 mPa.s (25.00 °C) | description viscosity 5738 mPa.s (25.00 °C) | description viscosity 4588 mPa.s (25.00 °C) | description viscosity 29.04 mPa.s (25.00 °C) |
| format mixture | format mixture | format mixture | format mixture |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| application(s) cleaning products | application(s) cleaning products | application(s) cleaning products | application(s) cleaning products |
| manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd | manufacturer/tradename Paragon Scientific Ltd |
| viscosity 7.520 mPa.s(25.00 °C) | viscosity 5738 mPa.s(25.00 °C) | viscosity 4588 mPa.s(25.00 °C) | viscosity 29.04 mPa.s(25.00 °C) |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Still not finding the right product?
Explore all of our products under Mineral Oil Rotational Viscosity Standard
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

