Skip to Content
Merck

Skip To

RTM5

Mineral Oil Rotational Viscosity Standard

viscosity 3.329 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
$321.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

$321.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

3.329 mPa.s(25.00 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Mineral Oil Rotational Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RTM31RTM30RTM17
description

viscosity 3.329 mPa.s (25.00 °C)

description

viscosity 5738 mPa.s (25.00 °C)

description

viscosity 4588 mPa.s (25.00 °C)

description

viscosity 250.0 mPa.s (25.00 °C)

format

mixture

format

mixture

format

mixture

format

mixture

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

viscosity

3.329 mPa.s(25.00 °C)

viscosity

5738 mPa.s(25.00 °C)

viscosity

4588 mPa.s(25.00 °C)

viscosity

250.0 mPa.s(25.00 °C)

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters