Skip to Content
Merck
All Photos(1)

Key Documents

RP-PP3-1

Recombinant Pru p 3.0102 (Prunus persica allergen 3.0102), peach

pkg of 250 μg, storage temp.:-20°C

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$1,180.00

$1,180.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
250 μG
$1,180.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.26

$1,180.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

grade

analytical standard

Assay

>95%

mol wt

10 kDa (under reducing conditions)

packaging

pkg of 250 μg

manufacturer/tradename

InBio

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

Source: nsLTP from peach expressed in Pichia pastoris
Formulation: Preservative and carrier-free in phosphate buffered saline, pH 7.4. Filtered through 0.22 μm filter.
Concentration: See product insert.
Purification: Purified from Pichia pastoris by multi-step HPLC. Purity > 95 % by silver stained SDS-PAGE.

Note:
(1) rPru p 3 has a C-terminal 6xHis-tag.
(2) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service