Skip to Content
Merck
All Photos(2)

Key Documents

RDD041

Sigma-Aldrich

Sodium chloride

Redi-Dri, ≥99%, DNase, RNase, and protease (none detected), free-flowing

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
≥99%
Pricing and availability is not currently available.

product line

Redi-Dri

Quality Level

Assay

≥99%

quality

free-flowing

pH

7

mp

801 °C (lit.)

foreign activity

DNase, RNase, and protease (none detected)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi Dri offers a more convenient and safer way to handle the products.

Features and Benefits

  • Redi-Dri packaging controls moisture for hygroscopic chemicals that commonly clump
  • Anhydrous powder remains free-flowing and easy to use
  • Safer to handle and weigh out for routine use in lab
  • Eliminates clumps, maintaining product moisture, quality, and attributes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service