Skip to Content
Merck
All Photos(1)

Key Documents

R8276

Sigma-Aldrich

Renin Substrate 1

>85% purity (HPLC), powder

Synonym(s):

Arg-Glu(EDANS)-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(DABCYL)-Arg

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,300.00

$1,300.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$1,300.00

About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

$1,300.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Product Name

Renin Substrate 1,

Assay

>85.0% (HPLC)

Quality Level

form

powder

mol wt

~2281

solubility

water: 1 mg/mL, clear, orange to red

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

dry ice

storage temp.

−20°C

General description

Substrate for human renin, an aspartic protease which plays an important role in blood-pressure regulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service