Skip to Content
Merck
All Photos(1)

Key Documents

PSRVD03

Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL

Nominal RI Value 1.5463 at 20 °C

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
$540.00

$540.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
30 ML
$540.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$540.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

General description

The product offers traceability of measurement to recognized national standards, and to units of measurement realized at the National Physical Laboratory (NPL) or other recognized national standards laboratories. Both density and refractive index values are fully traceable to National Institute Standards and Technology (NIST) and international protocols.

Application

The combined refractive index and density CRM (nominal RI value 1.5463 at 20 °C) is used to calibrate and verify refractometers such as handheld, Abbe and high-accuracy digital instruments.

Features and Benefits

  • CRM certified in strict adherence to ISO 17025 / ISO 17034 under UKAS accreditation
  • density measured and qualified in accordance with ASTM D1480
  • ensures full compliance to ASTM and IP test method protocols
  • low levels of uncertainty, guaranteeing maximum accuracy of data and dependable results
  • supplied in 30 mL volume sealed glass vials for ample measurement readings
  • 12-months shelf life as stated in the certificate of analysis (CoA)

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service