PHR8622
Palonosetron Related Compound C

certified reference material, pharmaceutical secondary standard
Synonym(s):
(3aS)-2-[(3R)-1-Azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benz[de]isoquinolin-1-one monohydrochloride, (3aS)-2-[(3R)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride
About This Item
grade
certified reference material
pharmaceutical secondary standard
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service