Skip to Content
Merck
All Photos(1)

Key Documents

PHR3891

(9β,11β,16α)-21-Chloro-9,11-epoxy-17-[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,20-dione

new

certified reference material

Synonym(s):

Mometasone Furoate Impurity D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29ClO6
CAS Number:
Molecular Weight:
484.97
UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

API family

mometasone

packaging

pkg of 30MG

application(s)

pharmaceutical

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service