PHR3874
Isobutyric acid
certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Methylpropionic acid
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
vapor density
3.04 (vs air)
vapor pressure
1.5 mmHg ( 20 °C)
autoignition temp.
824 °F
expl. lim.
10 %
packaging
pkg of 500MG
refractive index
n20/D 1.393 (lit.)
bp
153-154 °C (lit.)
mp
−47 °C (lit.)
density
0.95 g/mL at 25 °C (lit.)
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
CC(C)C(O)=O
InChI
1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)
InChI key
KQNPFQTWMSNSAP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
131.0 °F - closed cup
Flash Point(C)
55 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service