Skip to Content
Merck
All Photos(1)

Key Documents

PHR1950

Supelco

Spiramycin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Spiramycin, Formacidine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$303.00

$303.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 04, 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$303.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$303.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 04, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. S1100000

API family

spiramycin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

format

neat

storage temp.

-10 to -25°C

SMILES string

CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](OC2CCC(C(C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1OC3OC(C)C(OC4CC(C)(O)C(O)C(C)O4)C(C3O)N(C)C

InChI

1S/C43H74N2O14/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3/b13-12+,16-14+/t24-,25-,26?,27?,28?,29+,30?,31-,32+,34?,35?,36?,37?,38?,39+,40+,41?,42?,43?/m1/s1

InChI key

ACTOXUHEUCPTEW-JMRHEKERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Spiramycin belongs to the macrolide group of antibiotics,[1] widely used in the treatment of respiratory tract, skin and skin-structure, sexually transmitted, and various other infections.[2] Spiramycin is derived from the culture filtrate of Streptomyces ambofaciens[1] and its mode of action involves the exibition of antimicrobial activity by inhibiting ribosomal protein biosynthesis.[2]

Application

Spiramycin may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using liquid chromatography technique.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB1418 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation and determination of components of spiramycin in bulk powders and in pharmaceutical preparations by high-performance liquid chromatography
Bens AG, et al.
Chromatographia, 12(5), 294-298 (1979)
LC determination and stability assessment of macrolide antibiotics azithromycin and spiramycin in bulk and tablet samples
Mahmoudi A
International Letters of Natural Sciences, 47(5), 294-298 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service