Skip to Content
Merck
All Photos(1)

Key Documents

PHR1855

Supelco

Telmisartan

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Telmisartan, 4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$398.00

$398.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 09, 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$398.00

About This Item

Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$398.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 09, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000648
traceable to USP 1643419

API family

telmisartan

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)-c4ccccc4C(O)=O)-c5nc6ccccc6n5C

InChI

1S/C33H30N4O2/c1-4-9-30-35-31-21(2)18-24(32-34-27-12-7-8-13-28(27)36(32)3)19-29(31)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)

InChI key

RMMXLENWKUUMAY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Telmisartan belongs to the class of angiotensin II receptor antagonists (ARAs) used in the treatment of hypertension.

Application

This pharmaceutical secondary standard can also be used as follows:
  • Reversed-phase high-performance liquid chromatography (RP-HPLC) method-based multi-analysis of rosuvastatin, telmisartan, ezetimibe, and atorvastatin in pharmaceutical dosage formulations[1]
  • RP-HPLC analysis of telmisartan in the presence of its three impurities in its tablet formulation following an analytical quality-by-design (AQbD) approach[2]
  • Multi-determination of hydrochlorothiazide, amlodipine, and telmisartan by HPLC combined with spectrometric techniques without derivatization
  • Simultaneous estimation of telmisartan and hydrochlorothiazide by micellar high-pressure liquid chromatography and spectrophotometry-based methods in their combined tablet formulations[3]
  • RP-HPLC method-based determination of four cardiovascular drugs—atorvastatin, simvastatin, telmisartan, and irbesartan in bulk drugs and tablet dosage forms, following ICH guidelines

Biochem/physiol Actions

Telmisartan is a non-peptide AT1 angiotensin receptor antagonist.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3179 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemometric technique for the optimization of chromatographic system: Simultaneous HPLC determination of Rosuvastatin, Telmisartan, Ezetimibe and Atorvastatin used in combined cardiovascular therapy
Janardhanan VS, et al.
Arabian Journal of Chemistry, 9, S1378-S1387 (2016)
Assessment of the greenness of spectrophotometric and micellar liquid chromatographic methods using two approaches: Application to pharmaceutical analysis of hydrochlorothiazide and telmisartan
Ibrahim FA, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 148, 197-205 (2019)
AQbD-oriented development of a new LC method for simultaneous determination of telmisartan and its impurities
Dobricic V, et al.
Chromatographia, 80, 1199-1209 (2017)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service