Skip to Content
Merck
All Photos(1)

Documents

PHR1534

Supelco

Griseofulvin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Griseofulvin, (2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
Beilstein:
95226
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 180
traceable to Ph. Eur. G0600000
traceable to USP 1299007

API family

griseofulvin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

ClC1=C(O[C@@]2(C(OC)=CC(C[C@H]2C)=O)C3=O)C3=C(OC)C=C1OC

InChI

1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1

InChI key

DDUHZTYCFQRHIY-RBHXEPJQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Griseofulvin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3652 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

High?perfrmance liquid chromatographic analysis of griseofulvin in drug substance and solid dosage forms: Separation of impurities and metabolites.
Townley E and Roden P
Journal of Pharmaceutical Sciences, 69(5), 523-526 (1980)
Oil-in-water microemulsion high performance liquid chromatographic analysis of pharmaceuticals.
Marsh A, et al.
Chromatographia, 59(9), 531-542 (2004)
Damir Elmar Zecevic et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 87(2), 264-270 (2014-04-29)
Many upcoming drug candidates are pH-dependent poorly soluble weak bases in the pH range of the gastrointestinal tract. This often leads to a high in vivo variability and bioavailability issues. Aiming to overcome these limitations, the design of solid dispersions
Mark Sacchetti
Journal of pharmaceutical sciences, 103(9), 2772-2783 (2013-12-12)
Pharmaceutical materials, crystalline and amorphous, sorb water from the atmosphere, which affects critical factors in the development of drugs, such as the selection of drug substance crystal form, compatibility with excipients, dosage form selection, packaging, and product shelf-life. It is
Dipy M Vasa et al.
Journal of pharmaceutical sciences, 103(9), 2911-2923 (2014-05-16)
Fifteen model drugs were quenched from 3:1 (w/w) mixtures with polyethylene glycol 4000 (PEG4000). The resulting solids were characterized using powder X-ray diffraction (PXRD), analysis of pair distribution function-transformed PXRD data (where appropriate), hot-stage polarized light microscopy, and differential scanning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service