Skip to Content
Merck
All Photos(1)

Key Documents

PHR1401

Supelco

Zinc Pyrithione

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Zinc Pyrithione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$199.00

$199.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$199.00

About This Item

Empirical Formula (Hill Notation):
C10H8N2O2S2Zn
Molecular Weight:
317.70
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$199.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

zinc pyrithione

CofA

current certificate can be downloaded

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Zinc Pyrithione is used for controlling a variety of fungal infections. It is used commercially in personal care products, including antidandruff shampoos and skin creams.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3707 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and facile spectrophotometric tool for quantification of zinc pyrithione (ZPT) in suspension
Das S and Khubdikar K
Chemical Data Collections, 19, 100175-100175 (2019)
Heather S Loring et al.
Bioorganic & medicinal chemistry, 28(18), 115644-115644 (2020-08-24)
Sterile Alpha and Toll Interleukin Receptor Motif-containing protein 1 (SARM1) is a key therapeutic target for diseases that exhibit Wallerian-like degeneration; Wallerian degeneration is characterized by degeneration of the axon distal to the site of injury. These diseases include traumatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service