Skip to Content
Merck
All Photos(1)

Key Documents

PHR1375

Supelco

Ketoprofen

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ketoprofen, KP, 2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$156.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 668
traceable to Ph. Eur. K2000000
traceable to USP 1356632

API family

ketoprofen

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Ketoprofen is an active pharmaceutical compound used as a non-steroidal anti-inflammatory drug (NSAID). It is essentially a non-narcotic drug extensively utilized for mild to moderate pain relief.[1][2]

Application

Ketoprofen may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography and spectrophotometric technique.[1][2][3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is selective for COX-1.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3470 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous HPLC determination of ketoprofen and its degradation products in the presence of preservatives in pharmaceuticals
Dvorak J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(3), 625-629 (2004)
Development and validation of a stability-indicating high-performance liquid chromatographic assay for ketoprofen topical penetrating gel
Bempong DK and Bhattacharyya L
Journal of Chromatography A, 1073(1-2), 341-346 (2005)
UV-spectrophotometric determination of ketoprofen and paraben in a gel preparation by partial least-squares calibration
Blanco M, et al.
Fresenius Journal of Analytical Chemistry, 357(7), 967-972 (1997)
F Jamali et al.
Clinical pharmacokinetics, 19(3), 197-217 (1990-09-01)
Ketoprofen, a potent nonsteroidal anti-inflammatory drug (NSAID) of the 2-arylpropionic acid class, has been used clinically for over 15 years in Europe, and has recently been introduced in the United States. Although it possesses a chiral centre, with only the
Fabíola Garavello Prezotti et al.
Carbohydrate polymers, 113, 286-295 (2014-09-27)
Gellan gum/pectin beads were prepared by ionotropic gelation, using Al(3+) as crosslinker. High yield (92.76%) and entrapment efficiency (52.22-88.78%) were reached. Beads exhibited high circularity (0.730-0.849) and size between 728.95 and 924.56 μm. Particle size and circularity was increased by

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service