Skip to Content
Merck
All Photos(1)

Documents

PHR1345

Supelco

Thymine(Zidovudine Related Compound C)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Thymine, 2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
117880
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1754532

API family

zidovudine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~320 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thymine (Zidovudine Related Compound C) is a process related impurity of antiretroviral drug zidovudine. Zidovudine belongs to the class of drugs known as nucleoside reverse transcriptase inhibitors and is generally used in the prevention and treatment of HIV/AIDS.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Thymine (Zidovudine Related Compound C) may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1097 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zidovudine
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3962-3964 (2017)
Antiretroviral drugs
Warnke D
Journal of Clinical Pharmacology, 47(12), 1570-1579 (2007)
Zidovudine
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 45(1), 4630-4630 (2013)
T R Waters et al.
Mutation research, 462(2-3), 137-147 (2000-04-18)
About 23% of mutations in hereditary human diseases and 24% of mutations in p53 in human cancers are G to A transitions at sites of cytosine methylation suggesting that these sites are either foci for DNA damage, or foci for
M Weinfeld et al.
Progress in nucleic acid research and molecular biology, 68, 139-149 (2001-09-14)
The toxic and mutagenic effects of ionizing radiation are believed to be caused by damage to cellular DNA. We have made use of a novel immunoassay for thymine glycol to examine the removal of this lesion from the DNA of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service