Skip to Content
Merck
All Photos(1)

Key Documents

PHR1220

Supelco

Diethanolamine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1192808

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

API family

diethanolamine

CofA

current certificate can be downloaded

autoignition temp.

689 °F

expl. lim.

10.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Diethanolamine is commonly used as a chemical intermediate and as a surface-active agent in agricultural products, pharmaceutical formulations and cosmetics.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Ion Mobility: TWCCSN2 value of 119.8 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N PHR1220 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3260 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ecotoxicological evaluation of diethanolamine using a battery of microbiotests
Zurita LJ, et al.
Toxicology in vitro, 19(7), 879-886 (2005)
William Conway et al.
Environmental science & technology, 46(13), 7422-7429 (2012-05-25)
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six
Honghui Yu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2902-2910 (2014-08-15)
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic
S H Zeisel et al.
The Biochemical journal, 232(2), 403-408 (1985-12-01)
An understanding of the biosynthesis and metabolism of dimethylamine (DMA) is important because it is a precursor of dimethylnitrosamine (nitroso-DMA). DMA is the major short-chain aliphatic amine in human and rat urine. DMA is formed from trimethylamine (TMA), which, in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service