Skip to Content
Merck
All Photos(1)

Key Documents

PHR1113

Supelco

Sodium L-lactate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$177.00

$177.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$177.00

About This Item

Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
Beilstein:
4567087
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E325
NACRES:
NA.24

$177.00


Estimated to ship onMarch 29, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1614308

API family

sodium lactate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

163-165 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].C[C@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1

InChI key

NGSFWBMYFKHRBD-DKWTVANSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Sodium L-lactate is an alkalinizing compound, currently being researched for its efficacy in the treatment of acidosis.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7822 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Lind et al.
International journal of microbiology, 2010, 430873-430873 (2011-02-19)
Dairy propionibacteria are widely used in starter cultures for Swiss type cheese. These bacteria can ferment glucose, lactic acid, and glycerol into propionic acid, acetic acid, and carbon dioxide. This research examined the antifungal effect of dairy propionibacteria when glycerol
Michael G Tordoff et al.
Chemical senses, 39(6), 515-528 (2014-05-23)
Genetic ablation of calcium homeostasis modulator 1 (CALHM1), which releases adenosine triphosphate from Type 2 taste cells, severely compromises the behavioral and electrophysiological responses to tastes detected by G protein-coupled receptors, such as sweet and bitter. However, the contribution of
Maarten van der Velden et al.
Malaria journal, 14, 76-76 (2015-04-18)
Membrane-associated ATP binding cassette (ABC) transport proteins hydrolyze ATP in order to translocate a broad spectrum of substrates, from single ions to macromolecules across membranes. In humans, members from this transport family have been linked to drug resistance phenotypes, e.g.
Chen Huang et al.
Oncology reports, 33(2), 875-884 (2014-12-09)
Solid tumors grow faster and need more glucose than normal tissue; however, due to poor angiogenesis and excessive growth, tumors remote from blood vessels are always under glucose starvation. Even so, cancer cells remain alive in vivo. Thus, making cancer
Sofía Arriarán et al.
PloS one, 10(3), e0119572-e0119572 (2015-03-06)
Female and male adult Wistar rats were fed standard chow or a simplified cafeteria diet for one month. Then, the rats were killed and the white adipose tissue (WAT) in four sites: perigonadal, retroperitoneal, mesenteric and subcutaneous (inguinal) were sampled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service