Skip to Content
Merck
All Photos(1)

Key Documents

PHL89700

Harmaline

phyproof® Reference Substance

Synonym(s):

1-Methyl-7-methoxy-3,4-dihydro-β-carboline, 3,4-Dihydroharmine, 4,9-Dihydro-7-methoxy-1-methyl-3H-pyrido[3,4-b]indole, Dihydroharmine, Harmalol methyl ether, Harmidine, NSC 407285

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$325.00

$325.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$325.00

About This Item

Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
Beilstein:
207310
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$325.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

drug control

stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada

mp

232-234 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1ccc2c3CCN=C(C)c3[nH]c2c1

InChI

1S/C13H14N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-4,7,15H,5-6H2,1-2H3

InChI key

RERZNCLIYCABFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

Reactant for preparation of:
  • Acylated harmalines via Friedel-Crafts reaction
  • Phenacyl derivatives of 1-methyl-7-methoxy-β-carbolines as central nervous system affectors
  • Palladium harmaline DMSO chloro complex as anti-tumor agent

Natural product scaffold in preparation of:
  • Trypanothione reductase inhibitors†

Biochem/physiol Actions

CNS stimulant; may act through NMDA receptors

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service